Please login to your online account to display your discounted pricing

Pyridine - D5, deuteration degree min 99.8% for NMR spectroscopy MagniSolv™, EMD Millipore™


Quantity 10mL
Packaging Septum ampoule

Manufacturer: emd millipore corporation  1074750010

 View more versions of this product

Catalog No. M1074750010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Pyridine - D5
CAS 7291-22-7
Odor Characteristic
Packaging Septum ampoule
Ignition Point 482°C (900°F)
Vapor Pressure 20 hPa at 20°C (68°F)
Viscosity 0.97mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Quantity 10mL
Assay Percent Range 99.8% min.
Color Colorless
Molecular Formula C5D5N
Formula Weight 84.13

Specifications

Chemical Name or Material Pyridine - D5
CAS 7291-22-7
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula C5D5N
Formula Weight 84.13