Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-d5, 99.5% (Isotopic), contains 0.05% v/v TMS

Chemical Identifiers

Chemical Name or Material Pyridine-d5
CAS 7291-22-7
Name Note Isotopic
Formula Weight 84.14
MDL Number MFCD00044639
UN Number UN1282
Beilstein 114377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4234206

  SDS
alfa aesar™
4234206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4234214

  SDS
alfa aesar™
4234214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.14
Chemical Name or Material Pyridine-d5
Density 1.05g/mL
Name Note Isotopic
Boiling Point 115°C
Melting Point −42°C
Physical Form Liquid
Assay 99.5%
Quantity 5g
Concentration Contains 0.05% v/v TMS
Flash Point 20°C (68°F)
Refractive Index 1.506
MDL Number MFCD00044639
CAS 7291-22-7
UN Number UN1282
Beilstein 114377
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Odor Unpleasant
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.