missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-d5, 100%(Isotopic)

Chemical Identifiers

CAS 7291-22-7
Formula Weight 84.14
MDL Number MFCD00044639
UN Number UN1282
Beilstein 114377
Chemical Name or Material Pyridine-d5, Isotopic
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42343KV SDS Alfa Aesar™
42343KV
10 each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA42343SA SDS Alfa Aesar™
42343SA
2 each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7291-22-7
MFCD00044639
114377
84.14
UN1282
Pyridine-d5, Isotopic

Specifications

7291-22-7
Prescored Ampules
115°C
Liquid
20°C (68°F)
84.14
UN1282
Pyridine-d5, Isotopic
Unpleasant
1.05g/mL
−42°C
10 each
1.506
MFCD00044639
114377
100%

Safety and Handling

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE