Please login to your online account to display your discounted pricing

Pyridine, Anhydrous, 99.8% min., DriSolv™, EMD Millipore

$125.63 - $286.03

Chemical Identifiers

Chemical Name or Material Pyridine, Anhydrous
Name Note DriSolv™
CAS 110-86-1
Physical Form Liquid
Assay Percent Range 99.8%, min.
Molecular Formula C5H5N
Formula Weight 79.1
Synonym Azabenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX20127

 
emd millipore corporation
PX20127
100mL Each for $125.63

MPX20126

 
emd millipore corporation
PX20126
1L Each for $286.03
Description & Specifications

Specifications

Formula Weight 79.1
Quantity 100mL
Color Clear
Physical Form Liquid
Chemical Name or Material Pyridine, Anhydrous
Name Note DriSolv™
Assay Percent Range 99.8%, min.
Molecular Formula C5H5N
CAS 110-86-1
Recommended Storage Room Temperature
Synonym Azabenzene
Safety and Handling

Room Temperature