missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Formula Weight 79.1
Chemical Name or Material Pyridine
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7180500 Avantor Performance
718004
Glass 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

110-86-1
79.1
ACS Reagent Grade
C5H5N
Pyridine

Specifications

79.1
C5H5N
110-86-1
ACS Reagent Grade
500mL
Glass
Pyridine