Please login to your online account to display your discounted pricing

Pyridine, ACS, 99.0% min, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Pyridine
CAS 110-86-1
Purity Grade ACS Grade
Molecular Formula C5H5N
Formula Weight 79.1
MDL Number MFCD00011732
UN Number UN1282
Beilstein 103233
Merck Index 14,7970
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA19378K2

 
alfa aesar
19378K2
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 79.1
Quantity 1L
Chemical Name or Material Pyridine
TSCA TSCA
Odor Unpleasant
Refractive Index 1.5100
Flash Point 20°C (68°F)
Boiling Point 115°C
Physical Form Liquid
Color Colorless to Pale Yellow
Melting Point −42°C
Assay ≥99.0%
Molecular Formula C5H5N
MDL Number MFCD00011732
CAS 110-86-1
UN Number UN1282
Beilstein 103233
Merck Index 14,7970
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H302-H312-H332;
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Density 0.978
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
RTECS Number UR8400000
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H302-H312-H332;
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.