missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine, 99.8%, for biochemistry, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC447900025

 View more versions of this product

Catalog No. AC447900025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pyridine
0.9780g/mL
1.5085 to 1.5105
0.0005% max
115.0°C to 116.0°C
2.5L
99.75% min. (GC)
15,8081
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
C1=CC=NC=C1
79.1
CHEBI:16227
99.8%
0.0005% max
17°C
0.978
0.95 mPa.s (20°C)
-42.0°C
110-86-1
C5H5N
azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1
Biochemistry

Chemical Identifiers

110-86-1
79.1
azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196
CHEBI:16227
C1=CC=NC=C1
C5H5N
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 203-809-9

Documents