Learn More
Pyridine, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 22905M1
Description
Pyridine is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyridine | |
110-86-1 | |
Colorless liquid | |
115°C | |
20°C (68°F) | |
Unpleasant | |
1.51 | |
C5H5N | |
23 hPa (17mm Hg) at 20°C | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1 |
°C to 42°C | |
Colorless | |
0.978 | |
4 x 1 L | |
482°C (900°F) | |
Liquid | |
>99.5% | |
MFCD00011732 | |
103233 | |
Miscible with water,alcohol,ether,pet ether,and oils. | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
HPLC |
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Safety and Handling
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H312+H332-H315-H319
DOTInformation : Hazard Class: 3, Packing Group: II
EINECSNumber : 203-809-9
RTECSNumber : UR8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only