Learn More
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL
Supplier: Thermo Scientific Chemicals 339420025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyridine | |
110-86-1 | |
0.9780g/mL | |
2.5 L | |
Authentic | |
Liquid | |
0.001 % max. | |
0.978 | |
MFCD00011732 | |
0.005% Max. | |
01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1 | |
Extra Dry |
-42.0°C | |
Colorless | |
115.0°C to 116.0°C | |
17°C | |
Glass Bottle | |
1.5085 to 1.5105 | |
99.5% | |
C5H5N | |
0.95 mPa.s (20°C) | |
20, 181 | |
15, 8081 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
99.5% |
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Safety and Handling
GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only