missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine, 99.0% min., ACS, Alfa Aesar™

Maximum level of impurities: Evaporation residue 0.002%, H2O 0.1%, Cl 0.001%, SO4 0.001%, NH3 0.002%, Cu 5ppm, Reducing substances P.T., Solubility in water P.T.

Manufacturer:  Alfa Aesar 19378AP

 View more versions of this product

Catalog No. AA19378AP



Specifications

Specifications

Pyridine
110-86-1
Colorless liquid
C5H5N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
C1=CC=NC=C1
79.102
CHEBI:16227
Liquid
Unpleasant
500mL
For ACS analysis
ACS
99.0% min.
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1
482°C (900°F)
Colorless

Chemical Identifiers

110-86-1
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
C5H5N
MFCD00011732
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
CHEBI:16227
C1=CC=NC=C1
Safety and Handling

Safety and Handling

H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 203-809-9

SDS Documents