missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine, 99.0%, Extra Dry, anhydrous, AcroSeal™, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 610440010

 View more versions of this product

Catalog No. AC610440010



Description

Description

Specifications

Specifications

Pyridine
Vapor Pressure: 20mbar at 20°C
C5H5N
20, 181
15, 8081
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1
Extra Dry
Pass Test
1L
110-86-1
99.0%
MFCD00011732
01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196
C1=CC=NC=C1
79.102
CHEBI:16227
≥99.0%
0.9780g/mL
50ppm max.

Chemical Identifiers

110-86-1
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
C5H5N
MFCD00011732
azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196
CHEBI:16227
C1=CC=NC=C1
Safety and Handling

Safety and Handling

GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-809-9

RTECSNumber : UR8400000

TSCA : TSCA

SDS Documents