Please login to your online account to display your discounted pricing

Pyridine, 99.0%, Extra Dry, anhydrous, AcroSeal™, ACROS Organics™

$244.06 - $244.06

Chemical Identifiers

Chemical Name or Material Pyridine
CAS 110-86-1
Purity Grade Extra Dry
Assay ≥99.0%
Assay Percent Range 99.0%
Molecular Formula C5 H5 N
Formula Weight 79.1
MDL Number MFCD00011732
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC610440010

  SDS
fisher scientific acros
610440010
1L Each for $244.06
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 79.1
Quantity 1L
EINECS Number 203-809-9
TSCA TSCA
Purity Grade Extra Dry
Density 0.9780g/mL
RTECS Number UR8400000
Beilstein 20, 181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15, 8081
Chemical Name or Material Pyridine
Additional Information Vapor Pressure: 20mbar at 20°C
Assay Percent Range 99.0%
MDL Number MFCD00011732
Identification Pass Test
Water 50ppm max.
Molecular Formula C5 H5 N
CAS 110-86-1
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.