missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-4-carboxaldehyde, 97%

$46.80 - $469.49

Chemical Identifiers

CAS 872-85-5
Molecular Formula C6H5NO
Formula Weight 107.11
MDL Number MFCD00006425
Beilstein 105342
Synonym Isonicotinaldehyde
Chemical Name or Material Pyridine-4-carboxaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1065614 SDS Alfa Aesar™
A1065614
25g Each for $46.80
AAA1065622 SDS Alfa Aesar™
A1065622
100g Each for $134.69
AAA1065636 SDS Alfa Aesar™
A1065636
500g Each for $469.49
Specifications

Description

Applications
Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

Solubility
Soluble in water (20 mg/ml at 20°C), ethanol and acetone.

Notes
It is air and light sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

872-85-5
107.11
105342
Pyridine-4-carboxaldehyde
C6H5NO
MFCD00006425
Isonicotinaldehyde

Specifications

75°C (167°F)
−4°C
872-85-5
107.11
105342
Pyridine-4-carboxaldehyde
198°C
25g
C6H5NO
MFCD00006425
Isonicotinaldehyde
97%

Safety and Handling

GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NR9400000

TSCA : TSCA

Recommended Storage : 2°C to 8°C