missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-4-boronic acid hydrate, ca 10-15% water

$73.89 - $227.81

Chemical Identifiers

CAS 1692-15-5
Molecular Formula C5H6BNO2·xH2O
Formula Weight 122.92 (Anhydrous)
MDL Number MFCD08061353
Beilstein 471944
Synonym 4-Pyridylboronic acid
Chemical Name or Material Pyridine-4-boronic acid hydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1517903 SDS Alfa Aesar™
L1517903
1g Each for $73.89
AAL1517906 SDS Alfa Aesar™
L1517906
5g Each for $227.81
Specifications Description & Specifications

Chemical Identifiers

1692-15-5
122.92 (Anhydrous)
471944
Pyridine-4-boronic acid hydrate
C5H6BNO2·xH2O
MFCD08061353
4-Pyridylboronic acid

Specifications

>300°C
1692-15-5
122.92 (Anhydrous)
471944
Pyridine-4-boronic acid hydrate
1g
C5H6BNO2·xH2O
MFCD08061353
4-Pyridylboronic acid
≈10 to 15% water

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C