missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Pyridine-4-boronic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
1692-15-5 |
Molecular Formula |
C5H6BNO2 |
Molecular Weight (g/mol) |
122.918 |
InChI Key |
QLULGIRFKAWHOJ-UHFFFAOYSA-N |
Synonym |
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid |
PubChem CID |
2734379 |
IUPAC Name |
pyridin-4-ylboronic acid |
SMILES |
B(C1=CC=NC=C1)(O)O |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
1692-15-5 |
|
122.918 |
|
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid |
|
pyridin-4-ylboronic acid |
|
C5H6BNO2 |
|
QLULGIRFKAWHOJ-UHFFFAOYSA-N |
|
2734379 |
|
B(C1=CC=NC=C1)(O)O |
Specifications
|
1692-15-5 |
|
Amber Glass Bottle |
|
1g |
|
QLULGIRFKAWHOJ-UHFFFAOYSA-N |
|
pyridin-4-ylboronic acid |
|
2734379 |
|
97% |
|
97% |
|
C5H6BNO2 |
|
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid |
|
B(C1=CC=NC=C1)(O)O |
|
122.918 |
|
122.92 |
|
Pyridin-4-ylboronic acid |
SDS
missing translation for 'documents'