missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine-3-sulfonyl Chloride, TCI America™

Manufacturer:  TCI America P19355G

 View more versions of this product

Catalog No. P19355G



Specifications

Specifications

Pyridine-3-sulfonyl Chloride
Yellow
16133-25-8
MFCD03452747
CDRNYKLYADJTMN-UHFFFAOYSA-N
pyridine-3-sulfonyl chloride
3164136
Clear Liquid-Slightly Cloudy Liquid at 20°C
136°C
5g
C5H4ClNO2S
3265
C1=CC(=CN=C1)S(=O)(=O)Cl
177.602
177.60
≥98.0% (GC,T)

Chemical Identifiers

16133-25-8
177.602
CDRNYKLYADJTMN-UHFFFAOYSA-N
pyridine-3-sulfonyl chloride
C5H4ClNO2S
MFCD03452747
3164136
C1=CC(=CN=C1)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents