Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3-sulfonic acid, 98+%

Chemical Identifiers

Chemical Name or Material Pyridine-3-sulfonic acid
CAS 636-73-7
Molecular Formula C5H5NO3S
Formula Weight 159.16
MDL Number MFCD00006381
UN Number UN2585
Beilstein 120822
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1310114

  SDS
alfa aesar™
A1310114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1310136

  SDS
alfa aesar™
A1310136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.16
Chemical Name or Material Pyridine-3-sulfonic acid
Molecular Formula C5H5NO3S
MDL Number MFCD00006381
CAS 636-73-7
UN Number UN2585
Beilstein 120822
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
Quantity 25g
Assay ≥98%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UT7920000
Melting Point ≈350°C (dec.)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.