Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3-carboxaldoxime, 98%

$114.51 - $114.51

Chemical Identifiers

Chemical Name or Material Pyridine-3-carboxaldoxime
CAS 1193-92-6
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD00006409
Synonym Pyridine-3-aldoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2567914

  SDS
alfa aesar™
B2567914
25g Each for $114.51

AAB2567922

  SDS
alfa aesar™
B2567922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Pyridine-3-carboxaldoxime
Melting Point 148° to 151°C
Quantity 25g
CAS 1193-92-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UT4060000
Assay 98%
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD00006409
Synonym Pyridine-3-aldoxime
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.