Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3-carboxaldehyde, 97%

Chemical Identifiers

Chemical Name or Material Pyridine-3-carboxaldehyde
CAS 500-22-1
Molecular Formula C6H5NO
Formula Weight 107.11
MDL Number MFCD00006382
UN Number UN2924
Beilstein 105343
Synonym Nicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1460718

 
alfa aesar™
A1460718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1460730

 
alfa aesar™
A1460730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 107.11
Chemical Name or Material Pyridine-3-carboxaldehyde
TSCA TSCA
Assay 97%
Molecular Formula C6H5NO
MDL Number MFCD00006382
CAS 500-22-1
UN Number UN2924
Beilstein 105343
Synonym Nicotinaldehyde
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
RTECS Number QS2980000
Flash Point 60°C (140°F)
Odor Characteristic
Boiling Point 95° to 97°C (15mmHg)
Melting Point ≈8°C
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause an allergic skin reaction.