missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3-boronic acid pinacol ester, 98%

$68.30 - $216.01

Chemical Identifiers

CAS 329214-79-1
Molecular Formula C11H16BNO2
Molecular Weight (g/mol) 205.064
MDL Number MFCD03084758
InChI Key XEMDFESAXKSEGI-UHFFFAOYSA-N
Synonym 2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 3-pyridineboronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pyridine-3-boronic acid pinacol ester, pyridine-3-boronic acid, pinacol ester
PubChem CID 2734656
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH52513MD
View Documents
Alfa Aesar™
H52513MD
250mg
Each for $68.30
 
AAH5251303
View Documents
Alfa Aesar™
H5251303
1g
Each for $216.01
 
Description

Description

Specifications

Chemical Identifiers

329214-79-1
205.064
XEMDFESAXKSEGI-UHFFFAOYSA-N
2734656
B1(OC(C(O1)(C)C)(C)C)C2=CN=CC=C2
C11H16BNO2
MFCD03084758
2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 3-pyridineboronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pyridine-3-boronic acid pinacol ester, pyridine-3-boronic acid, pinacol ester
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Specifications

77°C to 80°C
329214-79-1
MFCD03084758
XEMDFESAXKSEGI-UHFFFAOYSA-N
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
2734656
98%
250mg
C11H16BNO2
2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 3-pyridineboronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pyridine-3-boronic acid pinacol ester, pyridine-3-boronic acid, pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN=CC=C2
205.064
205.07
Pyridine-3-boronic acid pinacol ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS