missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3-boronic acid 1,3-propanediol ester, 98+%

$46.41 - $160.38

Chemical Identifiers

CAS 131534-65-1
Molecular Formula C8H10BNO2
Molecular Weight (g/mol) 162.983
MDL Number MFCD01075749
InChI Key ZUUZNOFTKGDLLN-UHFFFAOYSA-N
Synonym 3-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid 1,3-propanediol ester, pyridine-3-boronic acid 1,3-propanediol cyclic ester, 1,3-propanediol cyclic ester, 2-3-pyridyl-1,3,2-dioxaborinane, 3-1,3,2 dioxaborinan-2-yl-pyridine, pyridine-3-boronic acid 1,3-propanediol ester, 3-pyridineboronic acid glycol ester, 2-3-pyridyl-1,3,2-dioxaborane, 2-pyridin-3-yl-1,3,2-dioxaborinane
PubChem CID 4198005
IUPAC Name 3-(1,3,2-dioxaborinan-2-yl)pyridine
SMILES B1(OCCCO1)C2=CN=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1701006 View Documents Alfa Aesar™
L1701006
5g Each for $160.38
 
AAL1701003 View Documents Alfa Aesar™
L1701003
1g Each for $46.41
 
Specifications

Chemical Identifiers

131534-65-1
162.983
ZUUZNOFTKGDLLN-UHFFFAOYSA-N
4198005
B1(OCCCO1)C2=CN=CC=C2
C8H10BNO2
MFCD01075749
3-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid 1,3-propanediol ester, pyridine-3-boronic acid 1,3-propanediol cyclic ester, 1,3-propanediol cyclic ester, 2-3-pyridyl-1,3,2-dioxaborinane, 3-1,3,2 dioxaborinan-2-yl-pyridine, pyridine-3-boronic acid 1,3-propanediol ester, 3-pyridineboronic acid glycol ester, 2-3-pyridyl-1,3,2-dioxaborane, 2-pyridin-3-yl-1,3,2-dioxaborinane
3-(1,3,2-dioxaborinan-2-yl)pyridine

Specifications

100°C to 104°C
131534-65-1
MFCD01075749
3-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid 1,3-propanediol ester, pyridine-3-boronic acid 1,3-propanediol cyclic ester, 1,3-propanediol cyclic ester, 2-3-pyridyl-1,3,2-dioxaborinane, 3-1,3,2 dioxaborinan-2-yl-pyridine, pyridine-3-boronic acid 1,3-propanediol ester, 3-pyridineboronic acid glycol ester, 2-3-pyridyl-1,3,2-dioxaborane, 2-pyridin-3-yl-1,3,2-dioxaborinane
B1(OCCCO1)C2=CN=CC=C2
162.983
162.98
Pyridine-3-boronic acid 1,3-propanediol ester
5g
C8H10BNO2
6138748
ZUUZNOFTKGDLLN-UHFFFAOYSA-N
3-(1,3,2-dioxaborinan-2-yl)pyridine
4198005
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning