missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3-boronic acid 1,3-propanediol ester, 98+%

Manufacturer:  Alfa Aesar™ L1701003

 View more versions of this product

Catalog No. AAL1701003



Specifications

Specifications

Pyridine-3-boronic acid 1,3-propanediol ester
1g
C8H10BNO2
6138748
ZUUZNOFTKGDLLN-UHFFFAOYSA-N
3-(1,3,2-dioxaborinan-2-yl)pyridine
4198005
≥98%
100°C to 104°C
131534-65-1
MFCD01075749
1,3-propanediol cyclic ester
B1(OCCCO1)C2=CN=CC=C2
162.983
162.98

Chemical Identifiers

131534-65-1
162.983
ZUUZNOFTKGDLLN-UHFFFAOYSA-N
4198005
B1(OCCCO1)C2=CN=CC=C2
C8H10BNO2
MFCD01075749
1,3-propanediol cyclic ester
3-(1,3,2-dioxaborinan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents