missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3,5-dicarboxylic acid, 98%

Manufacturer:  Alfa Aesar™ A1154018

 View more versions of this product

Catalog No. AAA1154018



Specifications

Specifications

Pyridine-3,5-dicarboxylic acid
50g
98%
MFCD00006393
Dinicotinic acid
C1=C(C=NC=C1C(=O)O)C(=O)O
167.12
CHEBI:46875
~325°C (decomposition)
499-81-0
C7H5NO4
131640
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
pyridine-3,5-dicarboxylic acid
10366
167.12

Chemical Identifiers

499-81-0
167.12
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
10366
pyridine-3,5-dicarboxylic acid
C7H5NO4
MFCD00006393
Dinicotinic acid
CHEBI:46875
C1=C(C=NC=C1C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents