Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3,4-dicarboxylic acid, 98%

$59.51 - $212.46

Chemical Identifiers

Chemical Name or Material Pyridine-3,4-dicarboxylic acid
CAS 490-11-9
Molecular Formula C7H5NO4
Formula Weight 167.12
MDL Number MFCD00006392
Beilstein 137242
Merck Index 14,2283
Synonym Cinchomeronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1458009

  SDS
alfa aesar™
A1458009
10g Each for $59.51

AAA1458018

  SDS
alfa aesar™
A1458018
50g Each for $212.46
Description & Specifications

Specifications

Formula Weight 167.12
Melting Point ≈255°C (dec.)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 10g
Molecular Formula C7H5NO4
CAS 490-11-9
Merck Index 14,2283
Beilstein 137242
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Pyridine-3,4-dicarboxylic acid
MDL Number MFCD00006392
Synonym Cinchomeronic acid
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.