Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-3,4-dicarbonitrile, 98+%

$99.30 - $367.65

Chemical Identifiers

Chemical Name or Material Pyridine-3,4-dicarbonitrile
CAS 1633-44-9
Molecular Formula C7H3N3
Formula Weight 129.12
MDL Number MFCD00010744
Beilstein 472545
Synonym Cinchomeronitrile; 3,4-Dicyanopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0768703

  SDS
alfa aesar™
L0768703
1g Each for $99.30

AAL0768706

  SDS
alfa aesar™
L0768706
5g Each for $367.65
Description & Specifications

Specifications

Formula Weight 129.12
Chemical Name or Material Pyridine-3,4-dicarbonitrile
Assay ≥98%
Molecular Formula C7H3N3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
MDL Number MFCD00010744
CAS 1633-44-9
Beilstein 472545
Synonym Cinchomeronitrile; 3,4-Dicyanopyridine
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 78° to 82°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.