Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2-carboxaldoxime methochloride, 97%

$45.26 - $471.01

Chemical Identifiers

Chemical Name or Material Pyridine-2-carboxaldoxime methochloride
CAS 51-15-0
Molecular Formula C7H9ClN2O
Formula Weight 172.62
MDL Number MFCD00011981
Beilstein 4163981
Merck Index 14,7703
Synonym 2-PAM chloride; Pyridine-2-aldoxime methochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2389022

 
alfa aesar™
B2389022
100g Each for $471.01

AAB2389006

 
alfa aesar™
B2389006
5g Each for $45.26
Description & Specifications

Specifications

Formula Weight 172.62
Chemical Name or Material Pyridine-2-carboxaldoxime methochloride
Assay 97%
Molecular Formula C7H9ClN2O
MDL Number MFCD00011981
CAS 51-15-0
Beilstein 4163981
Merck Index 14,7703
Synonym 2-PAM chloride; Pyridine-2-aldoxime methochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UU4200000
Melting Point ≈223°C (dec.)
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.