Learn More
Pyridine-2-carboxaldoxime methochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2389014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyridine-2-carboxaldoxime methochloride | |
∼223°C (decomposition) | |
MFCD00011981 | |
4163981 | |
pralidoxime chloride, 1-methylpyridinium-2-aldoxime chloride, 2-pam-cl, 2-hydroxyimino methyl-1-methylpyridin-1-ium chloride, 2-hydroxyimino methyl-1-methylpyridinium chloride, 2-formyl 1-methyl pyridinium oxime chloride | |
Cl.CN1C=CC=C\C1=C/N=O | |
172.61 | |
172.62 |
51-15-0 | |
C7H9ClN2O | |
25 g | |
14,7703 | |
HIGSLXSBYYMVKI-UHDJGPCESA-N | |
[(Z)-(1-methylpyridin-2-ylidene)methyl]-oxoazanium;chloride | |
57428996 | |
97% |
Chemical Identifiers
51-15-0 | |
172.61 | |
HIGSLXSBYYMVKI-UHDJGPCESA-N | |
57428996 |
C7H9ClN2O | |
MFCD00011981 | |
pralidoxime chloride, 1-methylpyridinium-2-aldoxime chloride, 2-pam-cl, 2-hydroxyimino methyl-1-methylpyridin-1-ium chloride, 2-hydroxyimino methyl-1-methylpyridinium chloride, 2-formyl 1-methyl pyridinium oxime chloride | |
Cl.CN1C=CC=C\C1=C/N=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 200-080-9
RTECSNumber : UU4200000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only