Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2-carboxaldoxime, 99%

$81.22 - $961.31

Chemical Identifiers

Chemical Name or Material Pyridine-2-carboxaldoxime
CAS 873-69-8
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD00006354
Beilstein 109061
Synonym Pyridine-2-aldoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1671414

 
alfa aesar™
A1671414
25g Each for $81.22

AAA1671422

 
alfa aesar™
A1671422
100g Each for $232.04

AAA1671436

 
alfa aesar™
A1671436
500g Each for $961.31
Description & Specifications

Specifications

Formula Weight 122.13
Quantity 25g
Melting Point 110° to 112°C
RTECS Number TJ4180000
Assay 99%
Molecular Formula C6H6N2O
Chemical Name or Material Pyridine-2-carboxaldoxime
MDL Number MFCD00006354
Health Hazard 1 Danger
CAS 873-69-8
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Synonym Pyridine-2-aldoxime
Beilstein 109061
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.