missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1139114

 View more versions of this product

Catalog No. AAA1139114



Description

Description

Pyridine-2-carboxaldehyde, 99% is used as an Intermediate of Bisacodyl. It is also used as an intermediate in pharamceutical research, Intermediate in organicc synethesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2-carboxaldehyde, 99% is used as an Intermediate of Bisacodyl. It is also used as an intermediate in pharamceutical research, Intermediate in organicc synethesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C .Store under inert gas. Stable under recommended storage conditions.Incompatible to Strong bases, Strong oxidizing agents and acids.
Specifications

Specifications

Pyridine-2-carboxaldehyde
-21°C
6.5
68°C (154°F)
C6H5NO
MFCD00006290
UN2810
Air and light sensitive
It is soluble in water.
C1=CC=NC(=C1)C=O
107.112
CHEBI:73012
99%
1121-60-4
1.124
180°C to 181°C
Characteristic
1.536
25 g
105341
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde
CSDSSGBPEUDDEE-UHFFFAOYSA-N
pyridine-2-carbaldehyde
14273
107.11

Chemical Identifiers

1121-60-4
107.112
CSDSSGBPEUDDEE-UHFFFAOYSA-N
14273
pyridine-2-carbaldehyde
C6H5NO
MFCD00006290
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde
CHEBI:73012
C1=CC=NC(=C1)C=O
Safety and Handling

Safety and Handling

GHS H Statement
H331-H318-H227-H302-H315-H317-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P311-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H318-H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 214-333-6

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only