Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone, 98%

$62.97 - $226.01

Chemical Identifiers

Chemical Name or Material Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone
CAS 70421-66-8
Molecular Formula C12H10N4O2
Formula Weight 242.24
MDL Number MFCD00012100
Synonym Picolinaldehyde 4-nitrophenylhydrazone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2438006

  SDS
alfa aesar™
B2438006
5g Each for $62.97

AAB2438014

  SDS
alfa aesar™
B2438014
25g Each for $226.01
Description & Specifications

Specifications

Formula Weight 242.24
Chemical Name or Material Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone
Molecular Formula C12H10N4O2
CAS 70421-66-8
Synonym Picolinaldehyde 4-nitrophenylhydrazone
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Melting Point 256° to 259°C
MDL Number MFCD00012100
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.