Learn More
Pyridine-2-boronic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5331403
Description
Pyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.
Notes
Incompatible with strong oxidizing agents.
Specifications
Pyridine-2-boronic acid | |
>300°C | |
MFCD00151844 | |
2-pyridineboronic acid, pyridine-2-boronic acid, 2-pyridinylboronic acid, 2-pyridylboronic acid, pyridin-2-boronic acid, pyridylboronic acid, boronic acid, 2-pyridinyl, 2-pyridineboronicacid, 2-pyridinyl-boronic acid, pyridinylboronic acid | |
OB(O)C1=CC=CC=N1 | |
122.92 | |
122.92 |
197958-29-5 | |
C5H6BNO2 | |
1 g | |
UMLDUMMLRZFROX-UHFFFAOYSA-N | |
pyridin-2-ylboronic acid | |
2762745 | |
95% |
Chemical Identifiers
197958-29-5 | |
122.92 | |
UMLDUMMLRZFROX-UHFFFAOYSA-N | |
2762745 | |
OB(O)C1=CC=CC=N1 |
C5H6BNO2 | |
MFCD00151844 | |
2-pyridineboronic acid, pyridine-2-boronic acid, 2-pyridinylboronic acid, 2-pyridylboronic acid, pyridin-2-boronic acid, pyridylboronic acid, boronic acid, 2-pyridinyl, 2-pyridineboronicacid, 2-pyridinyl-boronic acid, pyridinylboronic acid | |
pyridin-2-ylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only