missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2,4-dicarboxylic acid, 98%, may cont. up to ca 10% water

$87.43 - $1,186.78

Chemical Identifiers

CAS 499-80-9
Molecular Formula C7H5NO4
Formula Weight 167.12
MDL Number MFCD00149413
Beilstein 131631
Synonym Lutidinic acid
Chemical Name or Material Pyridine-2,4-dicarboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1440906 SDS Alfa Aesar™
A1440906
5g Each for $87.43
AAA1440914 SDS Alfa Aesar™
A1440914
25g Each for $370.97
AAA1440922 SDS Alfa Aesar™
A1440922
100g Each for $1,186.78
Specifications

Description

Applications
It is an in vitro and in cell inhibitor, as well as a known inhibitor of the histone lysine demethylases. 2,4-Pyridinedicarboxylic acid has been used in a study to determine that ruthenium(II) complexes exert antimetastatic effects on several tumor cell lines in vitro, achieved mostly by the effect on cell adhesion, migration and angiogenesis. It blocks the activity of 2-OG oxygenases, which include certain lysine demethylases and a variety of hydroxylases (e.g., prolyl, collagen, lysyl). 2,3 2,4-PDCA inhibits several Jumonji domain-containing lysine demethylases when used at low micromolar concentrations. Through its effects on hydroxylases, including prolyl hydroxlase 1 (IC50 = 1.5 µM), It modulates hypoxia inducing factor turnover, collagen synthesis, and plant cell wall formation. It can inhibit zinc-dependent enzymes, like metallo-β-lactamase.8 2,4-PDCA also affects and is translocated by organic anion transporters.

Solubility
Soluble in water at 20°C 4.5 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

499-80-9
167.12
131631
Pyridine-2,4-dicarboxylic acid
C7H5NO4
MFCD00149413
Lutidinic acid

Specifications

≈245°C (dec.)
2.5 to 3.5
C7H5NO4
MFCD00149413
Lutidinic acid
98%, may cont. up to ≈10% water
5g
499-80-9
167.12
131631
Pyridine-2,4-dicarboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA