missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2,4-dicarbonitrile, 97%

$117.88 - $117.88

Chemical Identifiers

CAS 29181-50-8
Molecular Formula C7H3N3
Formula Weight 129.12
MDL Number MFCD00014627
Beilstein 115936
Synonym 2,4-Dicyanopyridine
Chemical Name or Material Pyridine-2,4-dicarbonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1316903 SDS Alfa Aesar™
A1316903
1g Each for $117.88
AAA13169MD SDS Alfa Aesar™
A13169MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

29181-50-8
129.12
115936
Pyridine-2,4-dicarbonitrile
C7H3N3
MFCD00014627
2,4-Dicyanopyridine

Specifications

87° to 92°C
29181-50-8
129.12
115936
Pyridine-2,4-dicarbonitrile
1g
C7H3N3
MFCD00014627
2,4-Dicyanopyridine
97%

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA