missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2,3-dicarboxylic acid, 99%

$33.22 - $85.88

Chemical Identifiers

CAS 89-00-9
Molecular Formula C7H5NO4
Formula Weight 167.12
MDL Number MFCD00006295
Beilstein 137110
Merck Index 14,8073
Synonym Quinolinic acid
Chemical Name or Material Pyridine-2,3-dicarboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1141414 SDS Alfa Aesar™
A1141414
25g Each for $33.22
AAA1141422 SDS Alfa Aesar™
A1141422
100g Each for $85.88
Specifications

Description

Applications
Quinolinic acid is an endogenous NMDA agonist. Quinolinic acid is a metabolite of tryptophan that acts as a putative NMDA receptor agonist. Quinolinic acid is an “excitotoxic” metabolite and an agonist of N-methyl-D-aspartate receptors. properties of KYNA raise the possibility of a functional link between KYNA and QUIN in the brain which may be of relevance for an understanding of human neurodegenerative disorders. Quinolinic acid is a potent endogenous excitant at amino acid receptors in CNS.

Solubility
Soluble in water (partly), alkalies, dilute aqueous base, DMSO, and alcohol (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

89-00-9
167.12
137110
Quinolinic acid
C7H5NO4
MFCD00006295
14,8073
Pyridine-2,3-dicarboxylic acid

Specifications

≈188°C (dec.)
89-00-9
167.12
137110
Quinolinic acid
99%
25g
C7H5NO4
MFCD00006295
14,8073
Pyridine-2,3-dicarboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : US7967250

TSCA : TSCA