missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridine-2,3-dicarboxylic acid, 99%

Manufacturer:  Alfa Aesar™ A1141422

 View more versions of this product

Catalog No. AAA1141422



Description

Description

Applications
Quinolinic acid is an endogenous NMDA agonist. Quinolinic acid is a metabolite of tryptophan that acts as a putative NMDA receptor agonist. Quinolinic acid is an “excitotoxic” metabolite and an agonist of N-methyl-D-aspartate receptors. properties of KYNA raise the possibility of a functional link between KYNA and QUIN in the brain which may be of relevance for an understanding of human neurodegenerative disorders. Quinolinic acid is a potent endogenous excitant at amino acid receptors in CNS.

Solubility
Soluble in water (partly), alkalies, dilute aqueous base, DMSO, and alcohol (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Pyridine-2,3-dicarboxylic acid
100g
99%
MFCD00006295
14,8073
GJAWHXHKYYXBSV-UHFFFAOYSA-N
pyridine-2,3-dicarboxylic acid
1066
167.12
~188°C (decomposition)
89-00-9
C7H5NO4
137110
Quinolinic acid
C1=CC(=C(N=C1)C(=O)O)C(=O)O
167.12
CHEBI:16675

Chemical Identifiers

89-00-9
167.12
GJAWHXHKYYXBSV-UHFFFAOYSA-N
1066
pyridine-2,3-dicarboxylic acid
C7H5NO4
MFCD00006295
Quinolinic acid
CHEBI:16675
C1=CC(=C(N=C1)C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : US7967250

TSCA : TSCA

SDS Documents