Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridazine-4-carboxylic acid, 97%

$131.09 - $426.42

Chemical Identifiers

Chemical Name or Material Pyridazine-4-carboxylic acid
CAS 50681-25-9
Molecular Formula C5H4N2O2
Formula Weight 124.1
MDL Number MFCD00011576
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2724603

  SDS
alfa aesar™
H2724603
1g Each for $131.09

AAH2724606

  SDS
alfa aesar™
H2724606
5g Each for $426.42
Description & Specifications

Specifications

Formula Weight 124.1
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD00011576
CAS 50681-25-9
Chemical Name or Material Pyridazine-4-carboxylic acid
Molecular Formula C5H4N2O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈244°C
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.