PYREXPLUS™ Reusable Tubes with Screw Caps
Protective PVC coating helps prevent glass from shattering
$841.60 - $841.60
Specifications
Closure Color Black
Closure Material Phenolic
Design Type Threaded
Material Borosilicate Glass
Shape Round Bottom
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Thread Size Capacity (Metric) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Thread Size Capacity (Metric) Length (Metric) Price Quantity  
15-327C Corning Incorporated
C6982520X
20mm 18-415 34mL 150mm Case of 96 Each for $841.60
Specifications
Description
  • Round-bottom culture tubes are fully autoclavable to 121°C (250°F) and resistant to thermal shock
  • Deep-form phenolic cap that resists effects of autoclaving and features a glued-in, white-rubber liner
Do not place over direct heat or flame. Do not heat above 121°C (250°F) moist heat or 110°C (230°F) dry heat.
Description & Specifications
Specifications
Black
Threaded
Round Bottom
No
Rubber
Phenolic
Borosilicate Glass
No
Marking Spot