missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREXPLUS™ Media/Solution Bottles

Heavy-duty PYREXPLUS™ round bottles have a protective PVC coating to help prevent glass from shattering and reduces spills

Manufacturer:  PYREXPLUS™ C61626500

 View more versions of this product

Catalog No. 06-423-3C



Description

Description

Heavy duty bottle which can be used for storage as well as mixing and sampling. A protective PVC coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. Bottle comes complete with autoclavable, one-piece, green colored, polypropylene plug seal cap with drip-free pouring ring. Glass bead indicates full capacity line. Teal enameled graduations and marking spot. Thread size: GL 45. Neck opening: 29 mm I.D.

Order Info

Various other caps fit this bottle and are available for separate purchase: Polypropylene colored caps in a choice of orange, green, purple, red, and light gray for ease of sorting and identification; Vented membrane cap with 0.22μm PTFE hydrophobic membrane; PBT caps with silicone septa for syringe sampling or introduction of reagents.

Specifications

Specifications

Polypropylene
Screw Cap
6.9 in.
Wide
17 oz., 500 mL
17 oz.
3.3 in.
4/Cs.
Non-sterile
100 to 400 mL
GL45
86 mm
176 mm
Cylindrical
500 mL
Pyrexplus
Borosilicate Glass
4/Pk.
Green
SDS
Documents
Product Certifications