missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREXPLUS™ Coated Squibb Separatory Funnel, PTFE Standard Stopcock, Glass Standard Taper Stopper

Protective PVC coating reduces spills and helps prevent glass from shattering

Manufacturer:  Corning Incorporated C664022L

 View more versions of this product

Catalog No. 10-438H


 

Specifications Description & Specifications

Specifications

67.6 oz.
16cm
Squibb Separatory Funnel
Glass
2000mL
19.5 in.
2.5 in.
Pyrex™
6.2 in.
For replacement glass standard taper stoppers
1 Case
38
67.6 oz., 2000mL
49.6cm
65mm
6
Accessories