Please login to your online account to display your discounted pricing
PYREX™ Parr Weighing Bottle With Short Length External Std. Taper Joint
Choose from a variety of tall, low-form, or regular weighing bottles
$536.83 - $536.83
Products
Catalog Number Mfr. No. Price Quantity    
10-462-880
corning incorporated
C16842024
Case of 12 Each for $536.83
Description & Specifications
  • Tall- and low-form bottles with standard taper joint openings accept a closed-bottom, hollow Pennyhead style stopper
  • Bottles with external standard taper joint openings have closure-style cap that fits over the externally ground body
  • PARR bottle is ideal for use in weighing small samples
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.