PYREX™ VISTA™ Narrow Mouth Erlenmeyer Flask with Heavy Duty Rim
Designed with uniform wall thickness to provide the proper balance between mechanical strength and thermal shock resistance. PYREX™ VISTA™ Narrow Mouth Erlenmeyer Flasks have a heavy duty rim and durable construction.
Manufacturer:  PYREX™ C70980250
 View more versions of this product
Catalog No. 07-250-090
Description
Description
Designed with uniform wall thickness to provide the proper balance between mechanical strength and thermal shock resistance. PYREX™ VISTA™ Narrow Mouth Erlenmeyer Flasks have a heavy duty rim and durable construction.
These narrow mouth Erlenmeyer flasks have uniform wall thickness which provide the proper balance between mechanical strength and thermal shock resistance. Approximate graduations are in durable blue enamel. An extra large blue marking space is also provided. The Erlenmeyer flasks are designed to ASTM E-1404 Type 1.
Compliance
Designed to ASTM™ E-1404 Type 1
Specifications
Specifications
Blue Enamel graduations; Top style: Beaded rim
Pyrex Glass
24/Cs.
Extra Large Blue Marking
Narrow
250 mL
Narrow Mouth
12/Pk.
No
Non-sterile