PYREX™ VISTA™ Class A Volumetric Flasks with Glass Standard Taper Stopper
Necks are tooled for standard taper glass stoppers
Manufacturer:  PYREX™ C706401L
 View more versions of this product
Catalog No. 07-250-106
Description
Description
Necks are tooled for standard taper glass stoppers
These volumetric flasks provide precise volume measurement. The necks are tooled for standard taper glass stoppers. The graduation line is sharp and permanent and large blue block letters make the inscription easy to read. These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E-694 for volumetric ware, ASTM E-542 for ­calibration of volumetric ware and ASTM E-288 for volumetric flasks.
Compliance
Manufactured to Class A tolerances as established by ASTM™ E-694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks
Specifications
Specifications
Glass
Pyrex Glass
2/Cs.
Yes
Flat
1 L
Long
1/Pk.
Class A