PYREX™ VISTA™ Short Neck Round Bottom Boiling Flask, Standard Taper Joint
Designed for great shock resistance
Manufacturer: PYREX™ C70320100
Description
These round bottom boiling flasks have full length outer standard taper joints, but with shorter necks. They are manufactured from Code 7740 borosilicate glass. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance. The round bottom boiling flasks are designed to ASTM E-1403. Adapter Cat. Nos. 8800, 8820, and 8825 are used to connect flasks with different size joints, thus allowing numerous combinations for diverse assemblies.
Compliance
Designed to ASTM E-1403
Specifications
Round | |
100 mL | |
Short | |
2/Pk. |
No | |
Borosilicate Glass | |
4/Cs. | |
24/40 |