PYREX™ VISTA™ Class B Volumetric Flasks with Polyethylene Snap-Cap
Capacity tolerances are twice those specified for Class A volumetric ware
Manufacturer: PYREX™ C705811L
Description
These Class B volumetric flasks have capacity tolerances are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue block letters make inscriptions easy to read. Snap caps are supplied with all sizes.
Specifications
Flat | |
To Contain | |
LDPE | |
Glass,Pyrex Glass | |
6/Cs. |
Snap Cap | |
Blue | |
1 L | |
Long | |
1/Pk. |