PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TC
Calibrated “To Contain”
Manufacturer:  PYREX™ C7002250
 View more versions of this product
Catalog No. 07-250-062
Description
Description
These Class A graduated cylinders are calibrated 'to contain' and have blue enamel graduations. The cylinder capacity is in accordance with ASTM E-1272. The 10 mL size has a funnel top. For bumper guard only, see Cat. No. 3066.
Specifications
Specifications
Hexagonal
Blue
Non-sterile
A
±0.25 mL
Borosilicate Glass
50 mL
Single
To Contain
Round
ASTM E1272, ASTM E542, ASTM E694
225 mm
Graduated Cylinder
24 mm
Funnel Top, Pourout
Autoclavable
SDS
Documents
Product Certifications
Accessories