PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TD
Calibrated “To Deliver”
Manufacturer:  PYREX™ C7002425
 View more versions of this product
Catalog No. 07-250-067
Description
Description
Calibrated “To Deliver”
These Class A graduated cylinders are calibrated 'To Deliver' and have blue enamel graduations. The cylinder capacity is in accordance with ASTM E-1272. The 10 mL size has a funnel top for easier filling.
Specifications
Specifications
Hexagonal
Blue
Non-sterile
194 mm
Graduated Cylinder
Borosilicate Glass
25 mL
Autoclavable
To Deliver
Round
ASTM E1272, ASTM E542, ASTM E694
±0.17 mL
1/Pk.
18 mm
Single
SDS
Documents
Accessories