Corning™ PYREX™ Vacuum-Type Distillation Traps
Separable, with standard taper joints
Supplier: Corning™ 772941
Description
With a full length outer joint on the outer tube and an inner joint on the inner tube. The top and side tubulations are the same O.D. as the center tube and have a length of 75 mm.
Specifications
Vacuum Trap | |
42 mm | |
2.8 in. |
Pyrex™ | |
45/50 |