Please login to your online account to display your discounted pricing
PYREX™ Vacuum-Type Distillation Traps
Separable, with standard taper joints
Manufacturer: corning incorporated  C772938
 View more versions of this product
Catalog No. 11-184-1C
  • / Case of 2 Each
Description & Specifications
Specifications
Diameter (English) Outer 1.50 in.
Diameter (Metric) Outer 38mm
Material Pyrex™
Product Type Vacuum Trap
Standard Taper Joint No. 40/50