PYREX™ Trypsinizing Flasks
Extra-deep baffles provide vigorous agitation
$235.21 - $742.68
Specifications
Material Borosilicate Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
09-552-42 Corning Incorporated
C446050
35mL Case of 3 Each for $235.21
09-552-46 Corning Incorporated
C44601L
700mL Case of 2 Each for $511.85
09-552-47 Corning Incorporated
C44602L
1500mL Case of 2 Each for $620.28
09-552-54 Corning Incorporated
C4465125
125mL Case of 3 Each for $733.66
09-552-55 Corning Incorporated
C4465250
250mL Case of 3 Each for $739.74
09-552-56 Corning Incorporated
C4465500
300mL Case of 3 Each for $742.68
Specifications
Description
  • Convert homogenous tissue samples into cell suspensions by digestion of connective proteins
  • With or without side arm
  • Side arm designs feature wells to prevent pour-out of unseparated tissues
  • Beaded neck flasks accept cotton plugs
  • Plain neck including flask side arm accommodates plastic closures (sold separately)
Description & Specifications
Specifications
Borosilicate Glass