missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Tall-Form Beakers

Graduated to indicate approximate content

Manufacturer:  Corning Incorporated C10601L

 View more versions of this product

Catalog No. 02-546G




Specifications

Specifications

33.81 oz.
1.53 in.
No
Yes
7.36 in.
Spout
Marking spot
White Enamel scale
Beaker
Borosilicate Glass
ASTM E960
8.9cm
Convenient for easy pouring
50 to 950mL
18.7cm
50mL
6 Pack
Berzelius
1000mL
Documents
Accessories